Assay Details
| Gene Symbol: NDUFAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010464.4 | XP_004010513.2 | 4 - 5 | 1014 | 59 | |
| XM_012181234.3 | XP_012036624.2 | 4 - 5 | 1018 | 59 | ||
More Information
Set Membership: |
|

NDUFAF1
Genomic Map