Assay Details
| Gene Symbol: CHRNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012654.4 | XP_004012703.1 | 4 - 5 | 381 | 60 | |
| XM_012185656.3 | XP_012041046.1 | 4 - 5 | 376 | 60 | ||
| XM_012185658.3 | XP_012041048.1 | 4 - 5 | 319 | 60 | ||
| XM_027974970.1 | XP_027830771.1 | 4 - 5 | 376 | 60 | ||
More Information
Set Membership: |
|

CHRNB1
Genomic Map