Assay Details
| Gene Symbol: FXYD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962306.1 | XP_027818107.1 | 5 - 6 | 267 | 63 | |
| XM_027962307.1 | XP_027818108.1 | 5 - 6 | 303 | 63 | ||
More Information
Set Membership: |
|

FXYD4
Genomic Map