Assay Details
| Gene Symbol: LOC101119610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021473.3 | XP_004021522.2 | 4 - 5 | 652 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101119610| Gene Symbol: LOC101119610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021473.3 | XP_004021522.2 | 4 - 5 | 652 | 76 | |
Set Membership: |
|