Assay Details
| Gene Symbol: ACOT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970565.1 | XP_027826366.1 | 6 - 7 | 1315 | 56 | |
| XM_027970570.1 | XP_027826371.1 | 6 - 7 | 943 | 56 | ||
| XM_027970574.1 | XP_027826375.1 | 6 - 7 | 848 | 56 | ||
More Information
Set Membership: |
|

ACOT11
Genomic Map