Assay Details
| Gene Symbol: LOC105612842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965950.1 | XP_027821751.1 | 4138 | 139 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105612842| Gene Symbol: LOC105612842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965950.1 | XP_027821751.1 | 4138 | 139 | ||
Set Membership: |
|