Assay Details
| Gene Symbol: CCNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970780.1 | XP_027826581.1 | 4 - 5 | 1442 | 70 | |
| GenBank mRNA | EU525164.1 | - | 1 - 2 | 190 | 70 | |
More Information
Set Membership: |
|

CCNA2
Genomic Map