Assay Details
| Gene Symbol: PCDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096111.2 | XP_014951597.2 | 1 - 2 | 3198 | 69 | |
| XM_027970352.1 | XP_027826153.1 | 1 - 2 | 3198 | 69 | ||
More Information
Set Membership: |
|

PCDH12
Genomic Map