Assay Details
| Gene Symbol: DTX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961212.1 | XP_027817013.1 | 6 - 7 | 1456 | 70 | |
| XM_027961213.1 | XP_027817014.1 | 7 - 8 | 1621 | 70 | ||
| XM_027961214.1 | XP_027817015.1 | 5 - 6 | 1315 | 70 | ||
| XM_027961215.1 | XP_027817016.1 | 6 - 7 | 1480 | 70 | ||
More Information
Set Membership: |
|

DTX2
Genomic Map