Assay Details
| Gene Symbol: OMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002018.4 | XP_004002067.1 | 6 - 7 | 1233 | 63 | |
| XM_012155871.3 | XP_012011261.1 | 6 - 7 | 1239 | 63 | ||
| XM_012155936.3 | XP_012011326.1 | 6 - 7 | 1177 | 63 | ||
| XM_027970658.1 | XP_027826459.1 | 6 - 7 | 1278 | 63 | ||
More Information
Set Membership: |
|

OMA1
Genomic Map