Assay Details
| Gene Symbol: GNGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012176340.3 | XP_012031730.1 | 3 - 4 | 277 | 95 | |
| XM_012176341.3 | XP_012031731.1 | 3 - 4 | 274 | 95 | ||
| XM_015095188.2 | XP_014950674.1 | 3 - 4 | 279 | 95 | ||
More Information
Set Membership: |
|

GNGT1
Genomic Map