Assay Details
| Gene Symbol: POLM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015095357.2 | XP_014950843.2 | 8 - 9 | 2123 | 61 | |
| XM_027968732.1 | XP_027824533.1 | 8 - 9 | 2124 | 61 | ||
| XM_027968733.1 | XP_027824534.1 | 9 - 10 | 1908 | 61 | ||
| XM_027968735.1 | XP_027824536.1 | 8 - 9 | 2124 | 61 | ||
| XM_027968736.1 | XP_027824537.1 | 8 - 9 | 1009 | 61 | ||
| XM_027968737.1 | XP_027824538.1 | 8 - 9 | 2123 | 61 | ||
| XM_027968738.1 | XP_027824539.1 | 8 - 9 | 1394 | 61 | ||
| XM_027968739.1 | XP_027824540.1 | 8 - 9 | 1227 | 61 | ||
More Information
Set Membership: |
|

POLM
Genomic Map