Assay Details
| Gene Symbol: YAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094733.2 | XP_014950219.2 | 3 - 4 | 494 | 72 | |
| XM_027967438.1 | XP_027823239.1 | 4 - 5 | 481 | 72 | ||
| XM_027967439.1 | XP_027823240.1 | 3 - 4 | 404 | 72 | ||
| XM_027967440.1 | XP_027823241.1 | 3 - 4 | 394 | 72 | ||
More Information
Set Membership: |
|

YAF2
Genomic Map