Assay Details
| Gene Symbol: LOC101105400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006568.4 | XP_004006617.1 | 4 - 5 | 841 | 77 | |
| XM_012174717.3 | XP_012030107.1 | 4 - 5 | 1003 | 77 | ||
More Information
Set Membership: |
|

LOC101105400
Genomic Map