Assay Details
| Gene Symbol: LOC114115245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970761.1 | XP_027826562.1 | 1 - 2 | 105 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027970761.1 transcripts.
Set Membership: |
|

LOC114115245
Genomic Map