Assay Details
| Gene Symbol: LOC114115245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970761.1 | XP_027826562.1 | 2 - 3 | 195 | 61 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114115245| Gene Symbol: LOC114115245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970761.1 | XP_027826562.1 | 2 - 3 | 195 | 61 | |