Assay Details
| Gene Symbol: SLC18A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964381.1 | XP_027820182.1 | 6 - 7 | 1027 | 58 | |
| XM_027964382.1 | XP_027820183.1 | 6 - 7 | 1027 | 58 | ||
More Information
Set Membership: |
|

SLC18A1
Genomic Map