Assay Details
| Gene Symbol: SRSF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971955.1 | XP_027827756.1 | 6 - 7 | 1380 | 70 | |
| XM_027971956.1 | XP_027827757.1 | 6 - 7 | 605 | 70 | ||
| XM_027971957.1 | XP_027827758.1 | 6 - 7 | 735 | 70 | ||
| XM_027971958.1 | XP_027827759.1 | 6 - 7 | 592 | 70 | ||
| XM_027971959.1 | XP_027827760.1 | 6 - 7 | 1383 | 70 | ||
More Information
Set Membership: |
|

SRSF5
Genomic Map