Assay Details
| Gene Symbol: ERAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012510.4 | XP_004012559.1 | 8 - 9 | 1129 | 61 | |
| XM_027975220.1 | XP_027831021.1 | 8 - 9 | 1134 | 61 | ||
| XM_027975221.1 | XP_027831022.1 | 7 - 8 | 1069 | 61 | ||
More Information
Set Membership: |
|

ERAL1
Genomic Map