Assay Details
| Gene Symbol: LOC101103610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012102994.3 | XP_011958384.1 | 3 - 4 | 326 | 78 | |
| XM_015103304.2 | XP_014958790.1 | 3 - 4 | 320 | 78 | ||
More Information
Set Membership: |
|

LOC101103610
Genomic Map