Assay Details
| Gene Symbol: NR1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104653.2 | XP_014960139.2 | 4 - 5 | 3212 | 73 | |
| XM_027962719.1 | XP_027818520.1 | 5 - 6 | 1201 | 73 | ||
More Information
Set Membership: |
|

NR1D2
Genomic Map