Assay Details
| Gene Symbol: ARHGEF33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094426.2 | XP_014949912.2 | 8 - 9 | 827 | 69 | |
| XM_027966912.1 | XP_027822713.1 | 8 - 9 | 827 | 69 | ||
| XM_027966913.1 | XP_027822714.1 | 8 - 9 | 827 | 69 | ||
| XM_027966914.1 | XP_027822715.1 | 8 - 9 | 827 | 69 | ||
More Information
Set Membership: |
|

ARHGEF33
Genomic Map