Assay Details
| Gene Symbol: RNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098516.2 | XP_014954002.2 | 4 - 5 | 2085 | 63 | |
| XM_027975274.1 | XP_027831075.1 | 4 - 5 | 2067 | 63 | ||
More Information
Set Membership: |
|

RNF135
Genomic Map