Assay Details
| Gene Symbol: LOC101122140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012099260.2 | XP_011954650.1 | 2 - 3 | 1421 | 82 | |
| XM_015102191.2 | XP_014957677.1 | 3 - 4 | 227 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101122140| Gene Symbol: LOC101122140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012099260.2 | XP_011954650.1 | 2 - 3 | 1421 | 82 | |
| XM_015102191.2 | XP_014957677.1 | 3 - 4 | 227 | 82 | ||