Assay Details
| Gene Symbol: SPOCK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104422.2 | XP_014959908.2 | 4 - 5 | 1313 | 72 | |
| XM_027962406.1 | XP_027818207.1 | 3 - 4 | 1310 | 72 | ||
More Information
Set Membership: |
|

SPOCK2
Genomic Map