Assay Details
| Gene Symbol: NR1I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012095158.3 | XP_011950548.2 | 2 - 3 | 313 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR1I2| Gene Symbol: NR1I2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012095158.3 | XP_011950548.2 | 2 - 3 | 313 | 54 | |
Set Membership: |
|