Assay Details
| Gene Symbol: LOC101120131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104196.2 | XP_011959586.1 | 10 - 11 | 1388 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101120131| Gene Symbol: LOC101120131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104196.2 | XP_011959586.1 | 10 - 11 | 1388 | 69 | |
Set Membership: |
|