Assay Details
| Gene Symbol: MID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022435.4 | XP_004022484.1 | 2 - 3 | 993 | 64 | |
| XM_012107586.3 | XP_011962976.1 | 2 - 3 | 991 | 64 | ||
| XM_027963528.1 | XP_027819329.1 | 2 - 3 | 767 | 64 | ||
More Information
Set Membership: |
|

MID2
Genomic Map