Assay Details
| Gene Symbol: LOC101120895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106207.3 | XP_011961597.3 | 4 - 5 | 347 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101120895| Gene Symbol: LOC101120895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106207.3 | XP_011961597.3 | 4 - 5 | 347 | 104 | |
Set Membership: |
|