Assay Details
| Gene Symbol: PTPN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017400.4 | XP_004017449.1 | 3 - 4 | 563 | 61 | |
| XM_012097887.2 | XP_011953277.1 | 3 - 4 | 532 | 61 | ||
| XM_012097888.3 | XP_011953278.1 | 3 - 4 | 493 | 61 | ||
More Information
Set Membership: |
|

PTPN11
Genomic Map