Assay Details
| Gene Symbol: MAP3K11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959582.1 | XP_027815383.1 | 2 - 3 | 1821 | 64 | |
| XM_027959583.1 | XP_027815384.1 | 2 - 3 | 1855 | 64 | ||
More Information
Set Membership: |
|

MAP3K11
Genomic Map