Assay Details
| Gene Symbol: OSBPL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002992.3 | XP_004003041.1 | 4 - 5 | 1260 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OSBPL11| Gene Symbol: OSBPL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002992.3 | XP_004003041.1 | 4 - 5 | 1260 | 64 | |
Set Membership: |
|