Assay Details
| Gene Symbol: RAB5IF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977164.1 | XP_027832965.1 | 3 - 4 | 593 | 76 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB5IF| Gene Symbol: RAB5IF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977164.1 | XP_027832965.1 | 3 - 4 | 593 | 76 | |