Assay Details
| Gene Symbol: TNFSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013707.4 | XP_004013756.1 | 2 - 3 | 202 | 110 | |
| XM_012187290.3 | XP_012042680.1 | 2 - 3 | 441 | 110 | ||
More Information
Set Membership: |
|

TNFSF4
Genomic Map