Assay Details
| Gene Symbol: LOC101103165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012185735.3 | XP_012041125.3 | 32 - 33 | 4964 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101103165| Gene Symbol: LOC101103165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012185735.3 | XP_012041125.3 | 32 - 33 | 4964 | 73 | |