Assay Details
| Gene Symbol: LOC101117891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960671.1 | XP_027816472.1 | 5 - 6 | 1205 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117891| Gene Symbol: LOC101117891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960671.1 | XP_027816472.1 | 5 - 6 | 1205 | 78 | |
Set Membership: |
|