Assay Details
| Gene Symbol: LOC101114349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012176323.3 | XP_012031713.2 | 5 - 6 | 1602 | 66 | |
| XM_027968445.1 | XP_027824246.1 | 4 - 5 | 1446 | 66 | ||
More Information
Set Membership: |
|

LOC101114349
Genomic Map