Assay Details
| Gene Symbol: MBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970216.1 | XP_027826017.1 | 2 - 3 | 274 | 65 | |
| XM_027970217.1 | XP_027826018.1 | 2 - 3 | 274 | 65 | ||
| XM_027970218.1 | XP_027826019.1 | 2 - 3 | 349 | 65 | ||
More Information
Set Membership: |
|

MBD3
Genomic Map