Assay Details
| Gene Symbol: SLC31A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004003.4 | XP_004004052.1 | 3 - 4 | 336 | 67 | |
| XM_012112956.3 | XP_011968346.1 | 3 - 4 | 1192 | 67 | ||
More Information
Set Membership: |
|

SLC31A2
Genomic Map