Assay Details
| Gene Symbol: LOC114117335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976195.1 | XP_027831996.1 | 5 - 6 | 975 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114117335| Gene Symbol: LOC114117335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976195.1 | XP_027831996.1 | 5 - 6 | 975 | 67 | |
Set Membership: |
|