Assay Details
| Gene Symbol: MIA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017912.4 | XP_004017961.2 | 5 - 6 | 409 | 67 | |
| XM_012098849.3 | XP_011954239.2 | 10 - 11 | 2205 | 67 | ||
| XM_012098851.3 | XP_011954241.2 | 10 - 11 | 2205 | 67 | ||
| XM_012098858.3 | XP_011954248.2 | 5 - 6 | 409 | 67 | ||
More Information
Set Membership: |
|

MIA2
Genomic Map