Assay Details
Gene Symbol: C1H1orf162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002319.4 | XP_004002368.1 | 5 - 6 | 699 | 79 | |
XM_027973054.1 | XP_027828855.1 | 5 - 6 | 699 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
