Assay Details
| Gene Symbol: TP53INP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097918.2 | XP_014953404.1 | 3 - 4 | 475 | 68 | |
| XM_015097921.2 | XP_014953407.1 | 3 - 4 | 544 | 68 | ||
| XM_015097922.2 | XP_014953408.1 | 2 - 3 | 426 | 68 | ||
| XM_027973211.1 | XP_027829012.1 | 3 - 4 | 464 | 68 | ||
More Information
Set Membership: |
|

TP53INP1
Genomic Map