Assay Details
| Gene Symbol: TP53INP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097918.2 | XP_014953404.1 | 4 - 5 | 826 | 92 | |
| XM_015097921.2 | XP_014953407.1 | 4 - 5 | 895 | 92 | ||
| XM_015097922.2 | XP_014953408.1 | 3 - 4 | 777 | 92 | ||

TP53INP1
Genomic Map