Assay Details
| Gene Symbol: LOC101112602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958143.1 | XP_027813944.1 | 13 - 14 | 1908 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101112602| Gene Symbol: LOC101112602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958143.1 | XP_027813944.1 | 13 - 14 | 1908 | 65 | |
Set Membership: |
|