Assay Details
| Gene Symbol: SEPTIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961533.1 | XP_027817334.1 | 7 - 8 | 880 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEPTIN1| Gene Symbol: SEPTIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961533.1 | XP_027817334.1 | 7 - 8 | 880 | 81 | |
Set Membership: |
|