Assay Details
| Gene Symbol: LOC101121385 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092152.2 | XP_014947638.2 | 10 - 11 | 1256 | 84 | |
| XM_027964822.1 | XP_027820623.1 | 10 - 11 | 1250 | 84 | ||
More Information
Set Membership: |
|

LOC101121385
Genomic Map