Assay Details
| Gene Symbol: CACNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006392.4 | XP_004006441.2 | 8 - 9 | 824 | 59 | |
| XM_004006393.4 | XP_004006442.2 | 8 - 9 | 766 | 59 | ||
| XM_015094696.2 | XP_014950182.2 | 8 - 9 | 3265 | 59 | ||
| XM_027967388.1 | XP_027823189.1 | 7 - 8 | 4167 | 59 | ||
More Information
Set Membership: |
|

CACNB3
Genomic Map