Assay Details
| Gene Symbol: GPIHBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012183471.3 | XP_012038861.2 | 4 - 5 | 490 | 70 | |
| XM_027973445.1 | XP_027829246.1 | 3 - 4 | 981 | 70 | ||
More Information
Set Membership: |
|

GPIHBP1
Genomic Map