Assay Details
| Gene Symbol: CPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012184096.3 | XP_012039486.1 | 4 - 5 | 1319 | 61 | |
| XM_012184100.3 | XP_012039490.1 | 4 - 5 | 1319 | 61 | ||
| XM_027973198.1 | XP_027828999.1 | 4 - 5 | 928 | 61 | ||
| XM_027973199.1 | XP_027829000.1 | 4 - 5 | 1031 | 61 | ||
More Information
Set Membership: |
|

CPQ
Genomic Map